Skip to Content
Virus Filters
Viresolve® Pro Solution family of virus filters with Viresolve® Pro Devices (6 sizes) and Viresolve® Pro Shield (6 sizes) & Viresolve® Pro Shield H Prefilters (6 sizes)
Viral safety is a key concern in biomanufacturing and is essential to assuring drug safety for patients. For more than two decades, our Viresolve® Pro solutions have set the industry standard in viral safety providing reliable virus retention and cost-effective filtration operations. Viresolve® Pro Solutions provide flexible templates for virus filtration in monoclonal antibodies (mAbs) and recombinant protein production
Key Benefits and Features
Leveraging our global quality systems and the Emprove® Program, our Viresolve® Pro Solutions come with extensive documentation and regulatory support, facilitating your risk assessment and process control.
Schematic view of a virus filter membrane filtering out the viruses while letting the product through with several arrows showing the high flux.
High Flux and Capacity
Two layers of asymmetric polyethersulfone (PES) membrane that enables high flow rates.
Schematic bar chart, together with an arrow, showing an 50% increase in productivity
Effective Virus Retention
Reliable performance across a broad range of operating conditions with log reduction values (LRVs) ≥ 4.0 for small, non-enveloped parvovirus.
Schematic view of the Viresolve® Pro Solution with 6 virus filter sizes, showing the scalability
User-Friendly Scalable Solutions
Simple installation, operation and integrity testing from pilot to full-scale manufacturing.
Virus Filtration Solutions for Typical and More Challenging Molecules
Viresolve® Pro Solutions provide flexible templates for virus filtration in monoclonal antibodies (mAbs) and recombinant protein production and deliver effective virus retention, high flux and capacity to improve your process efficiency. These solutions include membrane prefilters to remove protein aggregates that reduce virus filter capacity.
Viresolve® Pro Solution Filters; all 6 sizes from the largest-magnus 2.2 device-to the smallest-micro 40 device.
Typical mAbs
Viresolve® Pro Solution includes the Viresolve® Pro Device and adsorptive prefilters, Viresolve® Pro Shield and Viresolve® Pro Shield H. This well-established filtration solution provides effective virus retention, high flux and capacity, and consistent batch-to-batch performance for most mAbs and recombinant proteins across process development, clearance studies, and commercial manufacturing.
  • Viresolve® Pro Device: Virus retentive filter. 100% integrity tested during manufacturing
  • Viresolve® Pro Shield Prefilter: Surface modified membrane prefilter with cation-exchange adsorptive chemistry
  • Viresolve® Pro Shield H Prefilter: Surface modified membrane prefilter with mixed-mode adsorptive chemistry
More Challenging Molecules
Viresolve® Pro-S Solution is an advanced filtration solution designed for challenging process fluids, including hydrophobic or high-concentration molecules, providing effective virus retention and enhanced processing. It includes the Viresolve® Pro-S Device and Viresolve® Pro-S Shield and Shield H prefilters for process development and clearance studies.
  • Viresolve® Pro-S Device: Virus retentive filter. 100% integrity tested during manufacturing
  • Viresolve® Pro-S Shield: surface modified membrane filter with cation-exchange adsorptive chemistry
  • Viresolve® Pro-S Shield H: surface modified membrane filter with mixed-mode adsorptive chemistry
Viresolve® Pro-S filters in micro 40 format: the device, the shield, and the shield H.
Reliable Virus Filtration
The Viresolve® Pro virus filtration solutions offer options for effective virus filtration of any mAb or recombinant protein molecule. Each of these solutions will provide the clearance capabilities you rely on to assure product safety. Further details listed below.
Simplifying Compliance
The Viresolve® Pro Solution is backed by the Emprove® Program – offering product-specific testing data, quality statements and regulatory information to support different stages of development and manufacturing. The Emprove® Program simplifies your compliance needs.
Process Development Service
Optimizing virus filtration involves assessing multiple parameters for reliable, efficient and cost-effective operations. We work side-by-side with engineers and manufacturers to streamline their filtration step. For help with process development, reach out to Tech Service.
Viral Clearance Services
At Bioreliance® Contract Testing our viral clearance studies are designed and executed by regulatory and virology experts ensuring compliance for IND and BLA studies across global facilities in Singapore, China, the U.S., and the UK. We minimize risk and accelerate your market readiness.
Frequently Asked Questions
Selecting which virus filter depends on both your molecule and process needs. For most standard mAbs, Viresolve® Pro Solution is the preferred option and will provide high virus retention and excellent filter flux and capacity for a broad range of molecules and operating conditions. For molecules that may have high concentration, hydrophobicity or other biochemical characteristics that are challenging for efficient virus filtration, Viresolve® Pro-S Solution would be the recommended option. Our process development engineers and MSAT teams are available to help you evaluate options and optimize your process.
No, the Viresolve® Pro Solution should be caustic sanitized – this includes both the Device and prefilters. Viresolve® Pro-S Solution is only available in gamma-irradiated presterilized formats. Although stable through pH 9 for standard bioprocessing applications, the low protein binding Viresolve® Pro-S surface chemistry is not caustic stable.
For a high concentration mAb we would recommend you evaluate Viresolve® Pro-S Solution first. Performance on any filter will depend on multiple factors including concentration, hydrophobicity, buffer characteristics, etc. Both filters will provide reliable and effective virus retention.
Yes, prefilters are generally recommended for any virus filtration operation and improve capacity and process consistency by reducing the impacts of batch-to-batch variability. Although the low protein binding chemistry of Viresolve® Pro-S Solution is less susceptible to adsorptive fouling, we still recommend using the Viresolve® Pro-S membrane prefilters to maximize filter capacity and maintain flux.
Two membrane prefilters are offered for each of the Viresolve® Pro and Viresolve® Pro-S Solutions: Viresolve® Pro Shield and Shield H prefilters for the Viresolve® Pro Device, and Viresolve® Pro-S Shield and Shield H prefilters for the Viresolve® Pro-S Device. These prefilters have different operating ranges so you can use these to easily select your prefilter based on your process characteristics.