MilliporeSigma
Potassium iodide AnhydroBeads™, −10 mesh, 99.998% trace metals basis

Potassium iodide

Potassium iodide AnhydroBeads™, −10 mesh, 99.998% trace metals basis
Synonyms:
Potassium iodide
Linear Formula:
KI
CAS Number:
Molecular Weight:
166.00
EC Number:

Products

Shipping
Product Category
Brand
Feature
Formula Weight
Melting Point (°C)
pH Value
Physical Form
Product Line
Purity
Agency
Quality Segment
Application
Special Grade
Technique

Products

Showing 1-20 of 22
Page 1 of 2
Compare
Product Number
Description
Pricing
Page 1 of 2