MilliporeSigma
Os EnCat® 40 extent of labeling: 0.3 mmol/g Os loading

Os EnCat® 40

Os EnCat® 40 extent of labeling: 0.3 mmol/g Os loading
Synonyms:
Osmium oxide, Osmium tetroxide, microencapsulated, Osmium(VIII) oxide
Empirical Formula (Hill Notation):
O4Os
CAS Number:
Molecular Weight:
254.23

Products

Shipping
Brand
Boiling Point (°C)
Color
Formula Weight
Melting Point (°C)
pH Value
Quality Segment
Application
Reagent Type
Core

Products

Showing 1-2 of 2
Compare
Product Number
Description
Pricing