MilliporeSigma
Sodium citrate, dihydrate EMPLURA®

Sodium citrate tribasic dihydrate

Sodium citrate, dihydrate EMPLURA®
Synonyms:
Citric acid trisodium salt dihydrate, Trisodium citrate dihydrate, Sodium citrate tribasic dihydrate
Linear Formula:
HOC(COONa)(CH2COONa)2 · 2H2O
CAS Number:
Molecular Weight:
294.10

Products

Shipping
Product Category
Brand
Feature
Formula Weight
Melting Point (°C)
pH Value
Physical Form
Product Line
Purity
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Technique

Product Comparison Guide

Use the product attributes below to configure the comparison table. (Select up to 3 total.)

Showing 1-19 of 19
Compare
Product Number
Description
Pricing