Skip to Content
MilliporeSigma
2-Aminothiazole 97%

2-Aminothiazole

2-Aminothiazole 97%
Synonym(s):
2-Thiazolamine, 2-Aminothiazole
Empirical Formula (Hill Notation):
C3H4N2S
CAS Number:
Molecular Weight:
100.14

Products

Shipping
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-2 of 2
Compare
Product Number
Description
Pricing