MilliporeSigma
2-Hexadecanol 99%

2-Hexadecanol

2-Hexadecanol 99%
Synonym(s):
Methyl tetradecyl carbinol
Linear Formula:
CH3(CH2)13CH(OH)CH3
CAS Number:
Molecular Weight:
242.44
EC Number:

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing