MilliporeSigma
Lithium aluminium hydride (powder) for synthesis

Lithium aluminum hydride

Lithium aluminium hydride (powder) for synthesis
Synonyms:
LAH, Lithium alanate, Lithium tetrahydroaluminate, Lithium aluminium hydride
Linear Formula:
LiAlH4
CAS Number:
Molecular Weight:
37.95

Products

Shipping
Brand
Feature
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Greener Alternative Principles
Greener Category
Reagent Type
Special Grade

Product Comparison Guide

Use the product attributes below to configure the comparison table. (Select up to 3 total.)

Showing 1-5 of 5