MilliporeSigma
Zinc foil, 0.5m coil, thickness 0.1mm, as rolled, 99.95+%

Zinc

Zinc foil, 0.5m coil, thickness 0.1mm, as rolled, 99.95+%
Synonym(s):
Zinc, Zinc 20 Mesh
Linear Formula:
Zn
CAS Number:
Molecular Weight:
65.39

Products

Shipping
Brand
Boiling Point (°C)
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Diameter (mm)
Length (m)
Element
Quality Segment
Showing 1-20 of 201
Page 1 of 11
Compare
Product Number
Description
Pricing
Page 1 of 11