MilliporeSigma
Hydroxylamine hydrochloride free-flowing, Redi-Dri™, ACS reagent, 98.0%

Hydroxylamine hydrochloride

Hydroxylamine hydrochloride free-flowing, Redi-Dri™, ACS reagent, 98.0%
Synonym(s):
Hydroxylammonium chloride
Linear Formula:
NH2OH · HCl
CAS Number:
Molecular Weight:
69.49

Products

Shipping
Product Category
Brand
Formula Weight
Melting Point (°C)
pH Value
Physical Form
Product Line
Purity
Agency
Quality Segment
Markush Class
Markush Group
Reaction Type
Reagent Type
Special Grade
Technique