MilliporeSigma
β-Nicotinamide adenine dinucleotide, reduced disodium salt hydrate ~98%, pkg of 100 mg

β-Nicotinamide adenine dinucleotide, reduced disodium salt hydrate

β-Nicotinamide adenine dinucleotide, reduced disodium salt hydrate ~98%, pkg of 100 mg
Synonym(s):
β-DPNH, β-NADH, DPNH, Diphosphopyridine nucleotide, reduced form, NADH, β-Nicotinamide adenine dinucleotide, reduced disodium salt hydrate, NADH, Disodium Salt, Dihydronicotinamide adenine dinucleotide disodium salt
Empirical Formula (Hill Notation):
C21H27N7Na2O14P2
CAS Number:
Molecular Weight:
709.40