MilliporeSigma
Glycine tested according to Ph. Eur.

Glycine

Glycine tested according to Ph. Eur.
Synonym(s):
Aminoacetic acid, Aminoethanoic acid, Glycocoll, Glycine
Linear Formula:
NH2CH2COOH
CAS Number:
Molecular Weight:
75.07
EC Number:

Products

Shipping
Product Category
Brand
Biological Source
Color
Feature
Formula Weight
Gene Alias
Melting Point (°C)
Physical Form
pK Value
Product Line
Purity
Agency
Quality Segment
Functional Group
Application
Markush Class
Markush Group
Amino Acid
Technique