MilliporeSigma
Dichloromethane Biosynthesis OmniSolv®

Dichloromethane

Dichloromethane Biosynthesis OmniSolv®
Synonyms:
Methylene chloride, Dichloromethane
Empirical Formula (Hill Notation):
CH2Cl2
CAS Number:
Molecular Weight:
84.93

Products

Shipping
Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Product Line
Purity
Agency
Quality Segment
Application
Special Grade
Technique
Showing 1-20 of 34
Page 1 of 2
Compare
Product Number
Description
Pricing
Page 1 of 2