MilliporeSigma
Glycerol

Glycerol

Glycerol
Synonyms:
1,2,3-Propanetriol, Glycerin, Glycerol
Linear Formula:
HOCH2CH(OH)CH2OH
CAS Number:
Molecular Weight:
92.09

Products

Shipping
Product Category
Brand
Biological Source
Boiling Point (°C)
Color
Feature
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Agency
Format
Quality Segment
Greener Alternative Principles
Greener Category
Application
Special Grade
Technique

Product Comparison Guide

Use the product attributes below to configure the comparison table. (Select up to 3 total.)

Showing 1-20 of 28
Page 1 of 2
Page 1 of 2
Compare
Product Number
Description
Pricing
Page 1 of 2
Page 1 of 2