MilliporeSigma
Acetic acid 100% for HPLC LiChropur™

Acetic acid 100%

Acetic acid 100% for HPLC LiChropur™
Synonyms:
Acetic acid, Glacial acetic acid
Linear Formula:
CH3CO2H
CAS Number:
Molecular Weight:
60.05

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Agency
Quality Segment
Markush Class
Markush Group
Technique

Product Comparison Guide

Use the product attributes below to configure the comparison table. (Select up to 3 total.)

Showing 1-2 of 2
Compare
Product Number
Description
Pricing