Skip to Content
MilliporeSigma
AG 1478 InSolution, ≥98%, inhibitor EGF receptor tyrosine kinase

AG 1478

AG 1478 InSolution, ≥98%, inhibitor EGF receptor tyrosine kinase
Synonym(s):
AG 1478, InSolution AG 1478
Empirical Formula (Hill Notation):
C16H14ClN3O2
CAS Number:
Molecular Weight:
315.75

Products

Product Category
Brand
Color
Formula Weight
Physical Form
Research Area
Quality Segment