MilliporeSigma
Agar Strips LAC-y Tryptic Soy Agar with Beta-Lactamase, Total count, sterile; irradiated, HYCON®

Agar Strips LAC-y

Agar Strips LAC-y Tryptic Soy Agar with Beta-Lactamase, Total count, sterile; irradiated, HYCON®
Synonym(s):
Agar Strips

Products

Product Category
Brand
Physical Form
Compatibility
Quality Segment
Application
Microorganism Suitability
Sterility
Sterilization Method
Showing 1-1 of 1