MilliporeSigma
Agar Strips TC Tryptic Soy Agar, Total count HYCON®

Agar Strips TC

Agar Strips TC Tryptic Soy Agar, Total count HYCON®
Synonym(s):
Agar Strips

Products

Product Category
Brand
Physical Form
Compatibility
Quality Segment
Application
Microorganism Suitability
Showing 1-1 of 1
Compare
Product Number
Description
Pricing