MilliporeSigma
Agar Strips TSM Tryptic Soy Agar, Total count, HYCON®, sterile; aseptically filled

Agar Strips TSM

Agar Strips TSM Tryptic Soy Agar, Total count, HYCON®, sterile; aseptically filled
Synonym(s):
Agar Strips

Products

Product Category
Brand
Physical Form
Compatibility
Quality Segment
Application
Microorganism Suitability
Sterility
Sterilization Method
Showing 1-1 of 1