Skip to Content
MilliporeSigma
Alcian blue solution for microscopy

Alcian Blue solution

Alcian blue solution for microscopy
Synonym(s):
Alcian Blue 8XG solution

Products

Shipping
Product Category
Brand
Color
Physical Form
Quality Segment
Application
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing