Skip to Content
MilliporeSigma
Amidosulfuric acid EMPLURA®

Amidosulfuric acid

Amidosulfuric acid EMPLURA®
Synonym(s):
Amidosulfuric acid
Empirical Formula (Hill Notation):
H3NO3S
CAS Number:
Molecular Weight:
97.09

Products

Product Category
Brand
Formula Weight
Physical Form
Product Line
Quality Segment
Technique
Showing 1-3 of 3
Compare
Product Number
Description
Pricing