MilliporeSigma
Ammonia solution 28-30% for analysis EMPARTA® ACS

Ammonia solution 28-30%

Ammonia solution 28-30% for analysis EMPARTA® ACS
Synonyms:
Ammonium hydroxide solution, Ammonia aqueous, Ammonia water
Linear Formula:
NH4OH
CAS Number:
Molecular Weight:
35.05

Products

Brand
Formula Weight
Physical Form
Product Line
Agency
Quality Segment

Products

Showing 1-2 of 2