Skip to Content
MilliporeSigma
Anti-ABCB1 antibody produced in rabbit Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Anti-ABCB1 antibody produced in rabbit

Anti-ABCB1 antibody produced in rabbit Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Synonym(s):
Anti-ATP-binding cassette sub-family B member 1, Anti-CD243 antigen, Anti-Multidrug resistance protein 1, Anti-P-glycoprotein 1, DKFZp686H0155, DNL1, DRNI, FLJ38093, FLJ44902, GP170, MDR1, P-GP, PGY1

Products

Product Category
Brand
Biological Source
Clonality
Feature
Gene Alias
Isotype
Physical Form
Product Line
Antibody Form
Research Area
Conjugate
Quality Segment
Post Translational Modification
Species Reactivity
Technique