Anti-GAPDH Antibody
Products
Product Category
Brand
Biological Source
Clonality
Gene Alias
Antibody Form
Quality Segment
Package Size
Post Translational Modification
Species Reactivity
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing