Skip to Content
MilliporeSigma
Anti-LDL-R antibody produced in chicken affinity isolated antibody, buffered aqueous solution

Anti-LDL-R antibody produced in chicken

Anti-LDL-R antibody produced in chicken affinity isolated antibody, buffered aqueous solution
Synonym(s):
Anti-FH, Anti-FHC, Anti-LDLCQ2, Anti-LDLR, Anti-Low density lipoprotein receptor

Products

Product Category
Brand
Biological Source
Clonality
Gene Alias
Physical Form
Antibody Form
Research Area
Conjugate
Quality Segment
Post Translational Modification
Species Reactivity
Technique
Showing 1-1 of 1