Skip to Content
MilliporeSigma
Anti-LSP1 antibody produced in rabbit Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Anti-LSP1 antibody produced in rabbit

Anti-LSP1 antibody produced in rabbit Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Synonym(s):
Anti-47 kDa actin-binding protein, Anti-52 kDa phosphoprotein, Anti-Lymphocyte-specific antigen WP34, Anti-Lymphocyte-specific protein 1, Anti-Protein pp52

Products

Product Category
Brand
Biological Source
Clonality
Feature
Gene Alias
Physical Form
Product Line
Antibody Form
Research Area
Conjugate
Quality Segment
Post Translational Modification
Species Reactivity
Technique