MilliporeSigma
Anti-MAP2 antibody, Mouse Monoclonal clone HM-2, purified from hybridoma cell culture

Anti-MAP2 antibody, Mouse Monoclonal

Anti-MAP2 antibody, Mouse Monoclonal clone HM-2, purified from hybridoma cell culture
Synonyms:
Monoclonal Anti-MAP2 antibody produced in mouse

Products

Shipping
Product Category
Brand
Biological Source
Clonality
Gene Alias
Isotype
Physical Form
Antibody Form
Research Area
Conjugate
Quality Segment
Package Size
Post Translational Modification
Species Reactivity
Technique

Products

Showing 1-1 of 1