
Anti-Progesterone Receptor antibody produced in rabbit

Synonym(s):
NR3C3, PGR, PR, PRGR, nuclear receptor 3C3
Products
Product Category
Brand
Biological Source
Clonality
Gene Alias
Physical Form
Antibody Form
Research Area
Conjugate
Post Translational Modification
Species Reactivity
Technique
Showing 1-3 of 3
Compare
Product Number
Description
Pricing