Skip to Content
MilliporeSigma
Anti-PSMA1 antibody produced in rabbit Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Anti-PSMA1 antibody produced in rabbit

Anti-PSMA1 antibody produced in rabbit Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Synonym(s):
Anti-HC2, Anti-MGC14542, Anti-MGC14575, Anti-MGC14751, Anti-MGC1667, Anti-MGC21459, Anti-MGC22853, Anti-MGC23915, Anti-NU, Anti-PROS30, Anti-Proteasome (prosome, macropain) subunit, α type, 1

Products

Product Category
Brand
Biological Source
Clonality
Feature
Gene Alias
Physical Form
Product Line
Antibody Form
Research Area
Conjugate
Quality Segment
Post Translational Modification
Species Reactivity
Technique