Skip to Content
MilliporeSigma
Anti-Sodium Channel, Pan antibody produced in rabbit affinity isolated antibody, lyophilized powder

Anti-Sodium Channel, Pan antibody produced in rabbit

Anti-Sodium Channel, Pan antibody produced in rabbit affinity isolated antibody, lyophilized powder
Synonym(s):
Anti-SP19, CIN5, HH1, SCN5A, Sodium channel protein, cardiac muscle α-subunit

Products

Product Category
Brand
Biological Source
Clonality
Gene Alias
Physical Form
Antibody Form
Research Area
Conjugate
Quality Segment
Post Translational Modification
Species Reactivity
Technique