
Anti-ZC3H12D antibody produced in rabbit

Synonym(s):
Anti-C6orf95, Anti-DJ281H8.1, Anti-MCPIP4, Anti-Zinc finger CCCH-type containing 12D, Anti-MCP-1-induced protein 4
Products
Product Category
Brand
Biological Source
Clonality
Feature
Gene Alias
Physical Form
Product Line
Antibody Form
Research Area
Conjugate
Post Translational Modification
Species Reactivity
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing