MilliporeSigma
Ascentis® Express RP-Amide, 2 μm UHPLC Column 2 μm particle size, L × I.D. 10 cm × 3 mm

Ascentis® Express RP-Amide, 2 μm UHPLC Column

Ascentis® Express RP-Amide, 2 μm UHPLC Column 2 μm particle size, L × I.D. 10 cm × 3 mm

Products

Product Category
Brand
pH Value
Product Line
Agency
USP Suitability
Separation Technique
ID (mm)
Length (cm)
Quality Segment
Application
Column Material
Package Size
Particle Size (µm)
Pore Size (Å)
Technique