MilliporeSigma
Ascentis® RP-Amide HPLC Column 5 μm particle size, L × I.D. 25 cm × 2.1 mm

Ascentis® RP-Amide HPLC Column

Ascentis® RP-Amide HPLC Column 5 μm particle size, L × I.D. 25 cm × 2.1 mm

Products

Product Category
Brand
pH Value
Product Line
Agency
USP Suitability
Separation Technique
ID (mm)
Length (cm)
Application
Column Material
Package Size
Particle Size (µm)
Pore Size (Å)
Technique

Products

Showing 1-17 of 17
Compare
Product Number
Description
Pricing