Skip to Content
MilliporeSigma
Atglistatin ≥98% (HPLC)

Atglistatin

Atglistatin ≥98% (HPLC)
Synonym(s):
3-(4′-(Dimethylamino)-[1,1′-biphenyl]-3-yl)-1,1-dimethylurea, ATGL Inhibitor, Atglistatin
Empirical Formula (Hill Notation):
C17H21N3O
CAS Number:
Molecular Weight:
283.37

Products