Skip to Content
US
EN
Back
2,3-Dihydrofuran
CAS Number:
1191-99-7
Molecular Weight:
70.09
Products
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
ALNH9A9E6664
2,3-Dihydrofuran,
Expand
View Pricing
APOH11A9D84A
2,3-Dihydrofuran,
Expand
View Pricing