Skip to Content
US
EN
Products
Applications
Services
Resources
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Lab Automation
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Documents
Learning Center
New
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
2-Methylpyrrolidine
CAS Number:
765-38-8
Molecular Weight:
85.15
Products
Brand
Aladdin Scientific
Combi-Blocks, Inc.
PharmaBlock (USA), Inc.
Purity
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
COMH039FD359
2-Methylpyrrolidine,
Expand
View Pricing
PBKH9C0D192B
2-Methylpyrrolidine,
Expand
View Pricing
ALNH9A9F77AC
2-Methylpyrrolidine,
Expand
View Pricing