Skip to Content
US
EN
Back
(2R,3R,4R,5R)-2,3,4,5,6-Pentahydroxyhexanal
CAS Number:
2595-97-3
Molecular Weight:
180.16
Products
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
XPIH9BD041EC
(2R,3R,4R,5R)-2,3,4,5,6-Pentahydroxyhexanal,
Expand
View Pricing
BL3H160B8D1A
(2R,3R,4R,5R)-2,3,4,5,6-Pentahydroxyhexanal,
Expand
View Pricing