Skip to Content
US
EN
Products
Applications
Services
Resources
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Lab Automation
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Documents
Learning Center
New
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Get up to
22% off
for Pi Day through March 27.
Save Now
Back
4-(Pyridin-4-ylmethyl)aniline
CAS Number:
27692-74-6
Molecular Weight:
184.24
Products
Brand
AiFChem (an XtalPi company)
ChemBridge Corporation
ChemScene LLC (Preferred Partner)
Purity
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
XPIH9BCF408E
4-(Pyridin-4-ylmethyl)aniline,
Expand
View Pricing
CH4238555772
4-(Pyridin-4-ylmethyl)aniline,
Expand
View Pricing
CIAH987F23C0
4-(Pyridin-4-ylmethyl)aniline,
Expand
View Pricing