
5-Chloro-N-(4-((S)-5-((5-chlorothiophene-2-carboxamido)methyl)-2-oxooxazolidin-3-yl)phenyl)-N-(2-(2-oxo-2-((((S)-2-oxo-3-(4-(3-oxomorpholino)phenyl)oxazolidin-5-yl)methyl)amino)ethoxy)ethyl)thiophene-2-carboxamide

CAS Number:
Molecular Weight:
871.78
Products
Brand
Purity
Showing 1-3 of 3
Compare
Product Number
Description
Pricing