Skip to Content
US
EN
Back
Dodecylamine
CAS Number:
124-22-1
Molecular Weight:
185.35
Products
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
ALNH9A9E68A0
Dodecylamine,
Expand
View Pricing
ALNH9A9E68A1
Dodecylamine,
Expand
View Pricing
CIAH987F0B12
Dodecylamine,
Expand
View Pricing