Skip to Content
US
EN
Back
Tributylammonium (hemipyrophosphate)
CAS Number:
5975-18-8
Molecular Weight:
363.33
Products
Filter Products
Showing 1-1 of 1
Compare
Product Number
Description
Pricing
CIAH9A70A856
Tributylammonium (hemipyrophosphate),
Expand
View Pricing