Benzalkonium Chloride 50% Solution
Synonym(s):
Benzalkonium chloride solution, Alkylbenzyldimethylammonium chloride
CAS Number:
Products
Shipping
Brand
Agency
Format
Quality Segment
Application
Package Size
API Family
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing