Skip to Content
MilliporeSigma
Blood typing Intermediates for Further Manufacturing Use (FFMU) - RHD (Rh) Specificities Anti-D from the clone TH-28, Human IgM, Intermediate for further manufacturing use

Blood typing Intermediates for Further Manufacturing Use (FFMU) - RHD (Rh) Specificities

Blood typing Intermediates for Further Manufacturing Use (FFMU) - RHD (Rh) Specificities Anti-D from the clone TH-28, Human IgM, Intermediate for further manufacturing use

Products

Brand
Biological Source
Clonality
Isotype
Physical Form
Antibody Form
Quality Segment
Package Size
Technique
Showing 1-9 of 9
Compare
Product Number
Description
Pricing