MilliporeSigma
Boc-D-Phe-OH ≥99.0% (sum of enantiomers, TLC)

Boc-D-Phe-OH

Boc-D-Phe-OH ≥99.0% (sum of enantiomers, TLC)
Synonym(s):
Boc-D-phenylalanine, Boc-D-Phe-OH
Empirical Formula (Hill Notation):
C14H19NO4
CAS Number:
Molecular Weight:
265.30

Products

Shipping
Product Category
Brand
Formula Weight
Physical Form
Product Line
Purity
Quality Segment
Application
Markush Class
Markush Group
Optical Isomer
Amino Acid
Reaction Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing