Skip to Content
MilliporeSigma
Calpeptin ≥98% (HPLC)

Calpeptin

Calpeptin ≥98% (HPLC)
Synonym(s):
N-Benzyloxycarbonyl-L-leucylnorieucinal, Z-Leu-Nle-CHO, Calpeptin
Empirical Formula (Hill Notation):
C20H30N2O4
CAS Number:
Molecular Weight:
362.46

Products

Product Category
Brand
Color
Formula Weight
Physical Form
Purity
Research Area
Quality Segment