
Cetrimide agar

Products
Shipping
Product Category
Brand
pH Value
Physical Form
Agency
Chemical Composition
Diameter (mm)
Volume (ml)
Quality Segment
Application
Microorganism Suitability
Package Size
Sterility
Sterilization Method
Technique
Showing 1-3 of 3
Compare
Product Number
Description
Pricing