Cholera Toxin from Vibrio cholerae
Synonym(s):
CTX, Cholera enterotoxin, Choleragen
CAS Number:
Products
Shipping
Product Category
Brand
Physical Form
Purity
Chemical Composition
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing