MilliporeSigma
Collagen from human placenta Bornstein and Traub Type IV, powder

Collagen from human placenta

Collagen from human placenta Bornstein and Traub Type IV, powder
Synonyms:
Collagen from human placenta
CAS Number:
EC Number:

Products

Shipping
Product Category
Brand
Biological Source
Gene Alias
Physical Form
Product Line
Tissue
Quality Segment
Application
Technique