Skip to Content
MilliporeSigma
Dorsomorphin ≥98% (HPLC)

Dorsomorphin

Dorsomorphin ≥98% (HPLC)
Synonym(s):
6-[4-(2-Piperidin-1-ylethoxy)phenyl]-3-pyridin-4-ylpyrazolo[1,5-a]pyrimidine, AMPK Inhibitor, Compound C, AMPK Inhibitor, Compound C, InSolution AMPK Inhibitor, Compound C
Empirical Formula (Hill Notation):
C24H25N5O
CAS Number:
Molecular Weight:
399.49

Products

Product Category
Brand
Color
Formula Weight
Physical Form
Purity
Research Area
Quality Segment