
Eliprodil

Synonym(s):
α-(4-Chlorophenyl)-4-[(4-fluorophenyl)methyl]-1-piperidineethanol, SL 820715, Eliprodil
Empirical Formula (Hill Notation):
C20H23ClFNO
CAS Number:
Molecular Weight:
347.85
Products
Product Category
Brand
Color
Formula Weight
Gene Alias
Physical Form
Purity
Research Area
Quality Segment
Showing 1-2 of 2
Compare
Product Number
Description
Pricing