MilliporeSigma
Ethanol absolute for analysis EMPARTA® ACS

Ethanol absolute

Ethanol absolute for analysis EMPARTA® ACS
Synonyms:
Ethanol, Ethyl alcohol
Linear Formula:
CH3CH2OH
CAS Number:
Molecular Weight:
46.07

Products

Product Category
Brand
Boiling Point (°C)
Feature
Formula Weight
Physical Form
Product Line
Format
Quality Segment
Greener Alternative Principles
Greener Category
Application
Markush Class
Markush Group
Special Grade

Product Comparison Guide

Use the product attributes below to configure the comparison table. (Select up to 3 total.)

Showing 1-1 of 1
Compare
Product Number
Description
Pricing